Merck
  • A novel mitosis-specific Cep215 domain interacts with Cep192 and phosphorylated Aurora A for organization of spindle poles.
A novel mitosis-specific Cep215 domain interacts with Cep192 and phosphorylated Aurora A for organization of spindle poles.
Journal of cell science (2020-12-31)
Ryoko Kuriyama, Cody R Fisher
ABSTRACT
The centrosome, which consists of centrioles and pericentriolar material (PCM), becomes mature and assembles mitotic spindles by increasing the number of microtubules (MTs) emanating from the PCM. Among the molecules involved in centrosome maturation, Cep192 and Aurora A (AurA, also known as AURKA) are primarily responsible for recruitment of γ-tubulin and MT nucleators, whereas pericentrin (PCNT) is required for PCM organization. However, the role of Cep215 (also known as CDK5RAP2) in centrosome maturation remains elusive. Cep215 possesses binding domains for γ-tubulin, PCNT and MT motors that transport acentrosomal MTs towards the centrosome. We identify a mitosis-specific centrosome-targeting domain of Cep215 (215N) that interacts with Cep192 and phosphorylated AurA (pAurA). Cep192 is essential for targeting 215N to centrosomes, and centrosomal localization of 215N and pAurA is mutually dependent. Cep215 has a relatively minor role in γ-tubulin recruitment to the mitotic centrosome. However, it has been shown previously that this protein is important for connecting mitotic centrosomes to spindle poles. Based on the results of rescue experiments using versions of Cep215 with different domain deletions, we conclude that Cep215 plays a role in maintaining the structural integrity of the spindle pole by providing a platform for the molecules involved in centrosome maturation.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)