Merck
  • Structural Basis for Potent Neutralization of Betacoronaviruses by Single-Domain Camelid Antibodies.
Structural Basis for Potent Neutralization of Betacoronaviruses by Single-Domain Camelid Antibodies.
Cell (2020-05-07)
Daniel Wrapp, Dorien De Vlieger, Kizzmekia S Corbett, Gretel M Torres, Nianshuang Wang, Wander Van Breedam, Kenny Roose, Loes van Schie, Markus Hoffmann, Stefan Pöhlmann, Barney S Graham, Nico Callewaert, Bert Schepens, Xavier Saelens, Jason S McLellan
ABSTRACT
Coronaviruses make use of a large envelope protein called spike (S) to engage host cell receptors and catalyze membrane fusion. Because of the vital role that these S proteins play, they represent a vulnerable target for the development of therapeutics. Here, we describe the isolation of single-domain antibodies (VHHs) from a llama immunized with prefusion-stabilized coronavirus spikes. These VHHs neutralize MERS-CoV or SARS-CoV-1 S pseudotyped viruses, respectively. Crystal structures of these VHHs bound to their respective viral targets reveal two distinct epitopes, but both VHHs interfere with receptor binding. We also show cross-reactivity between the SARS-CoV-1 S-directed VHH and SARS-CoV-2 S and demonstrate that this cross-reactive VHH neutralizes SARS-CoV-2 S pseudotyped viruses as a bivalent human IgG Fc-fusion. These data provide a molecular basis for the neutralization of pathogenic betacoronaviruses by VHHs and suggest that these molecules may serve as useful therapeutics during coronavirus outbreaks.
Sigma-Aldrich
Bovine Serum Albumin solution, 30%±2% in 0.85% sodium chloride, aseptically filled