- Characterization of Immune Cell Subsets of Tumor Infiltrating Lymphocytes in Brain Metastases.
Characterization of Immune Cell Subsets of Tumor Infiltrating Lymphocytes in Brain Metastases.
The heterogeneity of tumor infiltrating lymphocytes (TILs) is not well characterized in brain metastasis. To address this, we performed a targeted analysis of immune-cell subsets in brain metastasis tissues to test immunosuppressive routes involved in brain metastasis. We performed multiplex immunofluorescence (mIF), using commercially available validated antibodies on formalin-fixed paraffin embedded whole sections. We quantitated the subsets of immune-cells utilizing a targeted panel of proteins including PanCK, CD8, CD4, VISTA and IBA-1, and analyzed an average of 15,000 cells per sample. Classifying tumors as either high (>30%) or low (<30%) TILs, we found that increased TILs density correlated with survival. Phenotyping these TILs we found tumors with low TILs had significantly higher expression of the immune-checkpoint molecule VISTA in tumor cells (p < 0.01) as well as in their microenvironment (p < 0.001). Contrastingly, the tumors with high TILs displayed higher levels of microglia, as measured by IBA-1 expression. Low TILs-tumors displayed CD8+ T-cells that co-express VISTA (p < 0.01) significantly more compared to high TILs group, where CD8+cells significantly co-express IBA-11 (p < 0.05). These results were supported by RNA analysis of a publicly available, independent cohort. Our work contributes to a growing understanding of the immune surveillance escape routes active in brain metastasis.