Merck
  • MicroRNA-200b relieves LPS-induced inflammatory injury by targeting FUT4 in knee articular chondrocytes in vitro.
MicroRNA-200b relieves LPS-induced inflammatory injury by targeting FUT4 in knee articular chondrocytes in vitro.
Experimental and therapeutic medicine (2021-03-12)
Yintai Li, Suizhuan Wei, Zhongping Zhang
ABSTRACT
Osteoarthritis (OA), characterized by the degeneration of articular cartilage, is a major problem in aging populations, and cartilage chondrocytes have been indicated to serve a curial role in the progression of OA. MicroRNA-200b-3p (miR-200b) was preliminarily identified to participate in OA. However, its role and mechanism of action in injured chondrocytes in OA remain unclear to date. In the present study, lipopolysaccharide (LPS)-treated cells isolated from normal knee articular cartilage were used to mimic inflammatory injury of OA chondrocytes. Cell viability, apoptosis and inflammatory responses were detected using Cell Counting Kit-8, flow cytometry and enzyme-linked immunosorbent assay, respectively. The expression levels of miR-200b and fucosyltransferase-4 (FUT4) were measured by reverse transcription-quantitative PCR and western blotting. The association between miR-200b and FUT4 was verified using TargetScan software, dual-luciferase reporter assay and RNA immunoprecipitation. The results indicated that LPS treatment decreased cell viability of primary chondrocytes, and increased apoptosis rate and production of IL-1β, IL-6 and TNF-α. The expression level of miR-200b was downregulated, and that of FUT4 was upregulated in OA cartilage tissues and LPS-treated normal chondrocytes compared with normal cartilage tissues and chondrocytes. Overexpression of miR-200b via transfection with miR-200b mimic inhibited the apoptosis rate and reduced the levels of IL-1β, IL-6 and TNF-α in LPS-stimulated chondrocytes. However, the suppressive effect of miR-200b overexpression on the LPS-induced inflammatory injury in chondrocytes was reversed by the restoration of FUT4 levels. Notably, FUT4 was indicated to be a downstream target of miR-200b and was negatively regulated by miR-200b. Taken together, the results of the current study indicated that miR-200b protected chondrocytes from LPS-induced inflammatory injury in vitro by targeting FUT4. These findings revealed the miR-200b/FUT4 axis as a potential candidate to target the degeneration of cartilages, thereby inhibiting the progression of OA.
Sigma-Aldrich
Magna RIP® RNA-Binding Protein Immunoprecipitation Kit, RNA Immunoprecipitation (RIP) Kit containing all necessary reagents to perform 12 individual RNA-binding protein immunoprecipitation (RIP) reactions using protein A/G magnetic beads.
Sigma-Aldrich
RIPAb+ Ago2 - RIP Validated Antibody and Primer Set, ascites fluid, from mouse
Sigma-Aldrich
RIPAb+ IGF2 mRNA-binding protein 3 - RIP Validated Antibody and Primer Set, from rabbit, purified by affinity chromatography