Merck
  • Regioselective Radical Reaction of Monometallofullerene Y@C2v(9)-C82 With N-arylbenzamidine Mediated by Silver Carbonate.
Regioselective Radical Reaction of Monometallofullerene Y@C2v(9)-C82 With N-arylbenzamidine Mediated by Silver Carbonate.
Frontiers in chemistry (2020-11-17)
Jia Li, Pengyuan Yu, Peng Lai, Jiajun Zou, Zhe Liu, Xiuguang Yi, Wei Wang, Changwang Pan
ABSTRACT
A novel radical reaction of monometallofullerene Y@C2v(9)-C82 with N-arylbezamidine (1) is successfully conducted through catalysis with silver carbonate. The high-performance liquid chromatographic and mass spectrum results demonstrate that the reaction is highly regioselective to afford only one monoadduct (2) with an imidazoline group added on C82 cage, and computations through density functional theory reveal the addition group is attached to a specific [5, 6]-bond (C20-C76) near the Y atom. Furthermore, the analysis of prymidalization angle of the carbon atoms demonstrates the geometry of carbon cage is in favor of the regioselective formation of isomer (20, 76).
Sigma-Aldrich
Silver(I) carbonate, ≥99.9% trace metals basis