Merck
  • Very long chain fatty acid β-oxidation in astrocytes: contribution of the ABCD1-dependent and -independent pathways.
Very long chain fatty acid β-oxidation in astrocytes: contribution of the ABCD1-dependent and -independent pathways.
Biological & pharmaceutical bulletin (2012-11-06)
Masashi Morita, Saori Shinbo, Akiko Asahi, Tsuneo Imanaka
ABSTRACT
Very long chain fatty acid (VLCFA) metabolism in astrocytes is important for the maintenance of myelin structure in central nervous system. To analyze the contribution of the ABCD1-dependent and -independent pathways to VLCFA metabolism in astrocytes, we prepared human glioblastoma U87 cells with a silencing of ABCD1 and primary astrocytes from abcd1-deficient mice, and measured fatty acid β-oxidation in the presence or absence of a potent inhibitor of carnitine palmitoyltransferase I, 2-[5-(4-chlorophenyl)pentyl]oxirane-2-carboxylate (POCA). In U87 cells, C24:0 β-oxidation was decreased to ca. 70% of the control in the presence of POCA, and the activity was further decreased to ca. 20% by the silencing of ABCD1. In mouse primary astrocytes, C24:0 β-oxidation was also decreased to ca. 70% of the control in the presence of POCA. The C24:0 β-oxidation in Abcd1-deficient primary astrocytes was ca. 60% of the wild-type cells and the activity was further decreased to ca. 25% in the presence of POCA. Compared to human skin fibroblasts, in which VLCFA β-oxidation is not significantly inhibited by POCA, approximately one-third of the overall VLCFA β-oxidation was inhibited in both types of astrocytic cells. These results suggest that VLCFA is indeed β-oxidized in ABCD1-dependent pathway, but the ABCD1-independent peroxisomal and mitochondrial β-oxidation pathways significantly contribute to VLCFA β-oxidation in astrocytic cells.
Sigma-Aldrich
Anti-Adrenoleukodystrophy Protein Antibody, a.a. 495-648, clone 1AL-2B4, ascites fluid, clone 1AL-2B4, Chemicon®