Merck
Integrin clustering induces kinectin accumulation.
Journal of cell science (2002-04-26)
Huan Tran, Roumen Pankov, Simon D Tran, Brian Hampton, Wilson H Burgess, Kenneth M Yamada
ABSTRACT
Integrin receptors mediate the formation of adhesion complexes and play important roles in signal transduction from the extracellular matrix. Integrin-based adhesion complexes (IAC) contain proteins that link integrins to the cytoskeleton and recruit signaling molecules, including vinculin, paxillin, focal adhesion kinase, talin and alpha-actinin. In this study, we describe a approximately 160 kDa protein that is markedly enriched at IAC induced by clustering integrins with fibronectin-coated beads. Protein sequence analysis reveals that this approximately 160 kDa protein is kinectin. Kinectin is an integral membrane protein found in endoplasmic reticulum, and it serves as a receptor for the motor protein kinesin. Fibronectin-induced IAC sequestered over half of the total cellular content of kinectin within 20 minutes. In addition, two other ER-resident proteins, RAP [low-density lipoprotein receptor-related protein (LRP) receptor-associated protein] and calreticulin, were found to be clustered at IAC, whereas kinesin was not. Our results identify a novel class of constituents of IAC.