Merck
  • Marine natural compound cyclo(L-leucyl-L-prolyl) peptide inhibits migration of triple negative breast cancer cells by disrupting interaction of CD151 and EGFR signaling.
Marine natural compound cyclo(L-leucyl-L-prolyl) peptide inhibits migration of triple negative breast cancer cells by disrupting interaction of CD151 and EGFR signaling.
Chemico-biological interactions (2019-11-02)
Deepak Kgk, Seema Kumari, Shailender G, Rama Rao Malla
ABSTRACT
Cyclo (L-Leucyl-L-Prolyl) peptide/CLP is a marine natural metabolite and well recognized as an antimicrobial and antioxidant agent with limited studies on anticancer activity. The current study aims to determine the effect of CLP on migration and growth of triple negative breast cancer cell lines. The anti-growth potential was evaluated by MTT, BrdU and TUNEL assays; DNA damage by γH2AX and Dead green assays; antimigration activity by Boyden chamber invasion and wound healing assays. Interaction of CLP with CD151 was resolved by PatchDock. Effect of CLP on the expression of transmembrane CD151 was evaluated by cell-based ELISA assay. The interaction between CD151 and EGFR was predicted by using FireDoc Web server. Impact of CLP on the interaction of CD151 with EGFR was evaluated by co-immunoprecipitation assay. The effect of CLP on the cell cycle and its controlling proteins was determined by Western blotting. CLP reduced the viability of MDA-MB-231 and MDA-MB-468 TNBC cell lines but not human breast healthy epithelial cell line (MCF-12A) similar to eribulin, standard. CLP also inhibited proliferation; cell cycle and migration. It induced DNA strand breaks, DNA damage, and cell death. It showed the most favorable interactions with CD151 in in silico docking and significantly reduced the expression of membrane-bound CD151 proteins. FireDoc Web study predicted the association between CD151 and EGFR with -29.13 kcal/mol of binding energy. CLP reduced the interaction of CD151 with EGFR along with the expression of cyclin D, CDK4, PAK, RAC1, and P27kiP1. This study concludes that CLP suppresses growth and migration by attenuating cell cycle of TNBC cell lines via EGFR and CD151 signaling. Thus, exploring the EGFR and CD151 signaling pathway targeted by CLP may provide a new approach in the treatment of TNBC.