Merck
  • Iron chelation inhibits mTORC1 signaling involving activation of AMPK and REDD1/Bnip3 pathways.
Iron chelation inhibits mTORC1 signaling involving activation of AMPK and REDD1/Bnip3 pathways.
Oncogene (2020-06-17)
Chaowei Shang, Hongyu Zhou, Wang Liu, Tao Shen, Yan Luo, Shile Huang
ABSTRACT
The mammalian target of rapamycin (mTOR) functions as two complexes (mTORC1 and mTORC2), regulating cell growth and metabolism. Aberrant mTOR signaling occurs frequently in cancers, so mTOR has become an attractive target for cancer therapy. Iron chelators have emerged as promising anticancer agents. However, the mechanisms underlying the anticancer action of iron chelation are not fully understood. Particularly, reports on the effects of iron chelation on mTOR complexes are inconsistent or controversial. Here, we found that iron chelators consistently inhibited mTORC1 signaling, which was blocked by pretreatment with ferrous sulfate. Mechanistically, iron chelation-induced mTORC1 inhibition was not related to ROS induction, copper chelation, or PP2A activation. Instead, activation of AMPK pathway mainly and activation of both HIF-1/REDD1 and Bnip3 pathways partially contribute to iron chelation-induced mTORC1 inhibition. Our findings indicate that iron chelation inhibits mTORC1 via multiple pathways and iron is essential for mTORC1 activation.
Sigma-Aldrich
Dp44mT, ≥98% (HPLC)