Merck
  • Standards of care for treatment of recurrent glioblastoma--are we there yet?
Standards of care for treatment of recurrent glioblastoma--are we there yet?
Neuro-oncology (2012-11-09)
Michael Weller, Timothy Cloughesy, James R Perry, Wolfgang Wick
ABSTRACT
Newly diagnosed glioblastoma is now commonly treated with surgery, if feasible, or biopsy, followed by radiation plus concomitant and adjuvant temozolomide. The treatment of recurrent glioblastoma continues to be a moving target as new therapeutic principles enrich the standards of care for newly diagnosed disease. We reviewed PubMed and American Society of Clinical Oncology abstracts from January 2006 to January 2012 to identify clinical trials investigating the treatment of recurrent or progressive glioblastoma with nitrosoureas, temozolomide, bevacizumab, and/or combinations of these agents. At recurrence, a minority of patients are eligible for second surgery or reirradiation, based on appropriate patient selection. In temozolomide-pretreated patients, progression-free survival rates at 6 months of 20%-30% may be achieved either with nitrosoureas, temozolomide in various dosing regimens, or bevacizumab. Combination regimens among these agents or with other drugs have not produced evidence for superior activity but commonly produce more toxicity. More research is needed to better define patient profiles that predict benefit from the limited therapeutic options available after the current standard of care has failed.
Sigma-Aldrich
CT-2A-Luc Mouse Glioma Cell Line, CT-2A-Luc mouse glioma cell line is a valuable mouse model for therapeutic research on brain malignancies.
Sigma-Aldrich
CT-2A Mouse Glioma Cell Line, CT-2A mouse glioma cell line is a valuable mouse model for therapeutic research on brain malignancies.