Merck
  • Characterization of immortal cystic fibrosis tracheobronchial gland epithelial cells.
Characterization of immortal cystic fibrosis tracheobronchial gland epithelial cells.
Proceedings of the National Academy of Sciences of the United States of America (1992-06-01)
A L Cozens, M J Yezzi, L Chin, E M Simon, W E Finkbeiner, J A Wagner, D C Gruenert
ABSTRACT
Tracheobronchial glands were isolated and cultured from a patient with cystic fibrosis (CF). Cultured epithelial cells were transformed with pSVori-. All transformed cell lines express cytokeratin filaments and at early passages express the junctional complex molecule cell CAM 120/80, indicating their epithelial origin. Several gland cell lines express antigens that localize to secretory cells in vivo. Cl- transport measured by 36Cl efflux shows that CF gland epithelial cells, like CF surface airway and nasal polyp epithelial cells, are unable to respond to increases in intracellular cAMP. However, they do produce an increase in intracellular cAMP after treatment with isoproterenol or forskolin. One CF gland cell line shows increased intracellular calcium in response to a number of agents and increased Cl- efflux comparable to that observed in a non-CF airway surface epithelial cell line after addition of calcium ionophore. All cell lines express CF transmembrane conductance regulator mRNA, as measured by PCR amplification of first-strand cDNA. The CF tracheobronchial gland cell lines described here are compound heterozygotes, having a single copy of the delta F508 mutation.
Sigma-Aldrich
6CFSMEo- Human Cystic Fibrosis Submucosal Gland Epithelial Cell Line, 6CFSMEo- human CF submucosal gland epithelial cell line was derived from a cystic fibrosis patient who was compound heterozygote for the ΔF508 and Q2X CFTR mutations.
Sigma-Aldrich
CFSMEo- Human Cystic Fibrosis Submucosal Gland Epithelial Cell Line, CFSMEo- human CF submucosal gland epithelial cell line was derived from a cystic fibrosis patient who was compound heterozygote for the ΔF508 and Q2X CFTR mutations.