Merck
  • Contilisant, a Tetratarget Small Molecule for Alzheimer's Disease Therapy Combining Cholinesterase, Monoamine Oxidase Inhibition, and H3R Antagonism with S1R Agonism Profile.
Contilisant, a Tetratarget Small Molecule for Alzheimer's Disease Therapy Combining Cholinesterase, Monoamine Oxidase Inhibition, and H3R Antagonism with S1R Agonism Profile.
Journal of medicinal chemistry (2018-07-04)
Óscar M Bautista-Aguilera, Josiane Budni, Francielle Mina, Eduarda Behenck Medeiros, Winnie Deuther-Conrad, José M Entrena, Ignacio Moraleda, Isabel Iriepa, Francisco López-Muñoz, José Marco-Contelles
ABSTRACT
Contilisant, a permeable, antioxidant, and neuroprotectant agent, showing high nM affinity at H3R and excellent inhibition of the monoamine oxidases and cholinesterases, is an affine and selective S1R agonist in the nanomolar range, based on the binding affinity and functional experiment, a result confirmed by molecular modeling. In addition, contilisant significantly restores the cognitive deficit induced by Aβ1-42 in the radial maze assay in an in vivo Alzheimer's disease test, comparing very favorably with donepezil.
Sigma-Aldrich
ASS234, ≥98% (HPLC)