Merck
  • A novel small-molecule IAP antagonist, AZD5582, draws Mcl-1 down-regulation for induction of apoptosis through targeting of cIAP1 and XIAP in human pancreatic cancer.
A novel small-molecule IAP antagonist, AZD5582, draws Mcl-1 down-regulation for induction of apoptosis through targeting of cIAP1 and XIAP in human pancreatic cancer.
Oncotarget (2015-09-01)
Jai-Hee Moon, Jae-Sik Shin, Seung-Woo Hong, Soo-A Jung, Ih-Yeon Hwang, Jeong Hee Kim, Eun Kyoung Choi, Seung-Hee Ha, Jin-Sun Kim, Kyoung-Mok Kim, Dae-Won Hong, Daejin Kim, Yeong Seok Kim, Jeong Eun Kim, Kyu-Pyo Kim, Yong Sang Hong, Eun Kyung Choi, Jung Shin Lee, Maureen Hattersley, Dong-Hoon Jin, Tae Won Kim
ABSTRACT
Inhibitor of apoptosis proteins (IAPs) plays an important role in controlling cancer cell survival. IAPs have therefore attracted considerable attention as potential targets in anticancer therapy. In this study, we investigated the anti-tumor effect of AZD5582, a novel small-molecule IAP inhibitor, in human pancreatic cancer cells. Treating human pancreatic cancer cells with AZD5582 differentially induced apoptosis, dependent on the expression of p-Akt and p-XIAP. Moreover, the knockdown of endogenous Akt or XIAP via RNA interference in pancreatic cancer cells, which are resistant to AZD5582, resulted in increased sensitivity to AZD5582, whereas ectopically expressing Akt or XIAP led to resistance to AZD5582. Additionally, AZD5582 targeted cIAP1 to induce TNF-α-induced apoptosis. More importantly, AZD5582 induced a decrease of Mcl-1 protein, a member of the Bcl-2 family, but not that of Bcl-2 and Bcl-xL. Interestingly, ectopically expressing XIAP and cIAP1 inhibited the AZD5582-induced decrease of Mcl-1 protein, which suggests that AZD5582 elicits Mcl-1 decrease for apoptosis induction by targeting of XIAP and cIAP1. Taken together, these results indicate that sensitivity to AZD5582 is determined by p-Akt-inducible XIAP phosphorylation and by targeting cIAP1. Furthermore, Mcl-1 in pancreatic cancer may act as a potent marker to analyze the therapeutic effects of AZD5582.
Sigma-Aldrich
AZD5582, ≥98% (HPLC)