Merck
  • Ferrocene-Based Conjugated Microporous Polymers Derived from Yamamoto Coupling for Gas Storage and Dye Removal.
Ferrocene-Based Conjugated Microporous Polymers Derived from Yamamoto Coupling for Gas Storage and Dye Removal.
Polymers (2020-03-28)
Zhiqiang Tan, Huimin Su, Yiwen Guo, Huan Liu, Bo Liao, Abid Muhammad Amin, Qingquan Liu
ABSTRACT
Conjugated microporous polymers (CMPs) have conjugated skeleton and permanent porosity, and exhibit huge potential in developing novel functional materials for resolving the challenging energy and environment issues. Metal-containing CMPs often exhibited unique properties. In the present manuscript, ferrocene-based conjugated microporous polymers (FcCMPs) were designed and synthesized with 1,1'-dibromoferrocene and 5,10,15,20-Tetrakis(4- bromophenyl) porphyrin (FcCMP-1) or Tetra (p-bromophenyl) methane (FcCMP-2) as building units via Yamamoto coupling. FcCMPs were amorphous, and exhibited excellent thermal and physicochemical stability. The BET surface area of FcCMP-1 and FcCMP-2 was 638 m2/g and 422 m2/g, respectively. In comparison with FcCMP-2, FcCMP-1 displayed better gas storage capacity due to higher porosity. FcCMPs were also used as an adsorbent for removal of methyl violet from aqueous solution, and exhibited excellent adsorption properties due to the interaction between electron-rich conjugated structure of the polymers and methyl violet with cationic groups. Moreover, FcCMPs could be extracted and regenerated by an eluent and then re-used for high efficient removal of methyl violet.
Sigma-Aldrich
Allyl methyl sulfone, 96%
Sigma-Aldrich
1,1′-Dibromoferrocene, 97%