Merck
  • Cytokines, Angiogenesis, and Extracellular Matrix Degradation are Augmented by Oxidative Stress in Endometriosis.
Cytokines, Angiogenesis, and Extracellular Matrix Degradation are Augmented by Oxidative Stress in Endometriosis.
Annals of laboratory medicine (2020-04-21)
Amalesh Nanda, Thangapandi K, Priyanka Banerjee, Mainak Dutta, Tsering Wangdi, Pramod Sharma, Koel Chaudhury, Saikat Kumar Jana
ABSTRACT
The effect of the interplay among inflammation, angiogenesis, extracellular matrix (ECM) degradation, and oxidative stress (OS) on the pathogenesis of endometriosis remains unclear. Previously, we demonstrated the role of OS in endometriosis. Here, we performed a comprehensive investigation of several molecules involved in inflammation, angiogenesis, and ECM degradation in women with endometriosis to study their interplay with OS. Blood samples were collected from women with endometriosis (N=80), as well as from women with tubal factor infertility as controls (N=80). Interleukin (IL)-1β, tumor necrosis factor-alpha, interferon-gamma, transforming growth factor-beta, IL-4, -10, -2, -6, -8, vascular endothelial growth factor (VEGF), matrix metalloproteinase (MMP)-2, -9, tissue inhibitor of metalloproteinases (TIMP)-1, -2, and cyclooxygenase (COX)-2 levels in serum samples were measured using an ELISA. Nuclear factor kappa-light-chain-enhancer of activated B cells (NF-κB) in peripheral blood mononuclear cells was measured using flow cytometry. Cytokines, VEGF, MMPs, and COX-2 were significantly higher and TIMPs were significantly lower in patients with endometriosis. Multivariate statistical analysis indicated that IL-10 was the most significant variable capable of discriminating endometriosis samples from controls. Deregulation of NF-κB activation by OS affects the expression of various cytokines in endometriosis. Elevated cytokine levels further up-regulate IL-10, which subsequently activates the MMPs, leading to excessive ECM degradation and angiogenesis. Moreover, IL-10 emerged as the most important molecule involved in the pathogenesis of endometriosis. Measurement of these molecules may help in better management of the patients with endometriosis.
Product Description
Ficoll® Paque Plus, Cytiva 17-1440-02, pack of 6 × 100 mL