Merck
  • Design, synthesis and biological evaluation of 2-alkoxycarbonyl-3-anilinoindoles as a new class of potent inhibitors of tubulin polymerization.
Design, synthesis and biological evaluation of 2-alkoxycarbonyl-3-anilinoindoles as a new class of potent inhibitors of tubulin polymerization.
Bioorganic chemistry (2020-02-23)
Romeo Romagnoli, Filippo Prencipe, Paola Oliva, Maria Kimatrai Salvador, Andrea Brancale, Salvatore Ferla, Ernest Hamel, Giampietro Viola, Roberta Bortolozzi, Leentje Persoons, Jan Balzarini, Sandra Liekens, Dominique Schols
ABSTRACT
A new class of inhibitors of tubulin polymerization based on the 2-alkoxycarbonyl-3-(3',4',5'-trimethoxyanilino)indole molecular skeleton was synthesized and evaluated for antiproliferative activity, inhibition of tubulin polymerization and cell cycle effects. The results presented show that the methoxy substitution and location on the indole nucleus plays an important role in inhibition of cell growth, and the most favorable position for the substituent was at C-6. In addition, a small-size ester function (methoxy/ethoxycarbonyl) at the 2-position of the indole core was desirable. Also, analogues that were alkylated with methyl, ethyl or n-propyl groups or had a benzyl moiety on the N-1 indolic nitrogen retained activity equivalent to those observed in the parent N-1H analogues. The most promising compounds of the series were 2-methoxycarbonyl-3-(3',4'.5'-trimethoxyanilino)-5-methoxyindole 3f and 1-methyl-2-methoxycarbonyl-3-(3',4'.5'-trimethoxyanilino)-6-methoxy-indole 3w, both of which target tubulin at the colchicine site with antitubulin activities comparable to that of the reference compound combretastatin A-4.
Sigma-Aldrich
3,4,5-Trimethoxyaniline, 97%