Merck
  • Characterization and differentiation of boiled pork from Tibetan, Sanmenxia and Duroc × (Landrac × Yorkshire) pigs by volatiles profiling and chemometrics analysis.
Characterization and differentiation of boiled pork from Tibetan, Sanmenxia and Duroc × (Landrac × Yorkshire) pigs by volatiles profiling and chemometrics analysis.
Food research international (Ottawa, Ont.) (2020-03-12)
Dong Han, Chun-Hui Zhang, Marie-Laure Fauconnier, Si Mi
ABSTRACT
To characterize and differentiate boiled pork from three different pig breeds (Tibetan, Sanmenxia and Duroc × (Landrace × Yorkshire)), the volatile compounds in each were analysed by gas chromatography-olfactometry-mass spectrometry (GC-MS/O) and electronic nose (E-nose) combined with chemometrics analysis. In total, 61 volatile compounds were identified, among which 25 compounds were selected as odour-active compounds in boiled pork. Moreover, seven odour-active compounds (hexanal, nonanal, 1-octen-3-ol, dimethyl disulphide, heptanal, 2-pentylfuran and 2-ethylfuran) were the main contributors to the integral flavour of boiled pork due to their higher odour activity values (OAVs) ranging from 17.3 to 524.2. The odour-active compounds were examined by principal component analysis (PCA), agglomerative hierarchical clustering (AHC) and partial least squares-discriminant analysis (PLS-DA). The results showed that boiled pork from the three pig breeds could be clearly distinguished, and twelve odour-active compounds, including (E,E)-2,4-decadienal, ethyl hexanoate, dimethyl disulphide, hexanal, 2-acetylthiazole, (E)-2-nonenal, 1-octen-3-ol, (E,E)-2,4-nonadienal, heptanal, (E)-2-octen-1-ol, styrene and (E)-2-octenal, were determined as potential flavour markers for discrimination. This study indicated that GC-MS/O and E-nose with chemometrics analysis are feasible methods to characterize and discriminate boiled pork from three pig breeds.
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
2-Methyl-3-heptanone, 99%