Merck
  • Substrate specificity of a recombinant D-lyxose isomerase from Providencia stuartii for monosaccharides.
Substrate specificity of a recombinant D-lyxose isomerase from Providencia stuartii for monosaccharides.
Journal of bioscience and bioengineering (2010-06-15)
Hyun-Jung Kwon, Soo-Jin Yeom, Chang-Su Park, Deok-Kun Oh
ABSTRACT
The specific activity and catalytic efficiency (k(cat)/K(m)) of the recombinant putative protein from Providencia stuartii was the highest for D-lyxose among the aldose substrates, indicating that it is a D-lyxose isomerase. Gel filtration analysis suggested that the native enzyme is a dimer with a molecular mass of 44 kDa. The maximal activity for D-lyxose isomerization was observed at pH 7.5 and 45 degrees C in the presence of 1 mM Mn(2+). The enzyme exhibited high isomerization activity for aldose substrates with the C2 and C3 hydroxyl groups in the left-hand configuration, such as D-lyxose, D-mannose, L-ribose, D-talose, and L-allose (listed in decreasing order of activity). The enzyme exhibited the highest activity for D-xylulose among all pentoses and hexoses. Thus, D-lyxose was produced at 288 g/l from 500 g/l D-xylulose by D-lyxose isomerase at pH 7.5 and 45 degrees C for 2 h, with a conversion yield of 58% and a volumetric productivity of 144 g l(-1) h(-1). The observed k(cat)/K(m) (920 mM(-1) s(-1)) of P. stuartiid-lyxose isomerase for D-xylulose is higher than any of the k(cat)/K(m) values previously reported for sugar and sugar phosphate isomerases with monosaccharide substrates. These results suggest that the enzyme will be useful as an industrial producer of D-lyxose.
Sigma-Aldrich
D-(−)-Lyxose, 99%