Merck
Fractal SERS nanoprobes for multiplexed quantitative gene profiling.
Biosensors & bioelectronics (2020-03-17)
Xiwei Wang, Bingyi Liu, Mingshu Xiao, Yuxiu Zou, Wei Lai, Hao Pei, Md Fazle Alam, Weijia Zhang, Ying Wan, Li Li
ABSTRACT
Quantitative analysis is critical for biological and chemical sensing applications, yet still remains a great challenge in surface-enhanced Raman spectroscopy (SERS). Here we report the development of a novel fractal SERS nanoprobe with robust internal calibration standard and high multiplexing capability for ultrasensitive detection of DNA and microRNA. This fractal SERS nanoprobe consists of a solid Au core of ~13 nm, an inner hollow gap of ~1 nm, and a stellate outer shell. The inner hollow gap enables the embedding of Raman tags that can serve as a self-calibrating internal standard to effectively correct the fluctuations of samples and measuring conditions. The outer shell morphology is highly tunable, which provides distinct SERS enhancement and enables a reproducible quantitative measurement of nucleic acids down to femtomolar level. In addition, the flexibility of encoding crosstalk-free Raman tag molecules makes such SERS sensor particularly attractive for multiplexed bioassays. This technique is simple, reliable, and of wide applicability to various genomic screening and diagnostic applications.
Sigma-Aldrich
1,2-Di(4-pyridyl)ethylene, 97%