Merck
  • Modulatory effect of diclofenac on antispasmodic effect of pitofenone in cholinergic spasm.
Modulatory effect of diclofenac on antispasmodic effect of pitofenone in cholinergic spasm.
Indian journal of experimental biology (2004-07-21)
Shrinivas K Kulkarni, Chandrashekhar S Patil, Naveen K Jain, Amarjit Singh
ABSTRACT
Biliary, ureteric and intestinal colic are extremely common clinical conditions associated with smooth muscle spasm. In the present study, antispasmodic activity was carried out against acetylcholine (10-640 ng/ml)-induced contractions on guinea pig ileum. Acetylcholine (10-640 ng/ml) induced concentration-dependent contraction of smooth muscle. Diclofenac, in varying concentration (9.4 x 10(-5) mol/l and 14.1 x 10(-5) mol/l) shifted the concentration response curve of acetylcholine to the right without suppressing the maximal response. However, in higher concentration diclofenac (18.9 x 10(-5) mol/l) blocked the response in an unsurmountable fashion. Further, analgin (11.09 x 10(-5), 16.63 x 10(-5) and 22.18 x 10(-5) mol/l) in equimolar concentrations did not alter the concentration response curve of acetylcholine, but in higher concentration analgin (44.36 x 10(-5) mol/l) also blocked the response in an unsurmountable fashion. Pitofenone (2.5 x 10(-6) mol/l) also, shifted the concentration response curve of acetylcholine to right in a parallel fashion with no change in maximal response. The present study confirms the potent antispasmodic activity of diclofenac-pitofenone combination in comparison to analgin-pitofenone in molar equivalent concentration (in comparison to diclofenac) against acetylcholine-induced contractions of guinea pig ileum.
Sigma-Aldrich
Pitofenone hydrochloride, ≥98% (HPLC)