Merck
  • Biotransformation and toxicity effect of monoanthraquinone dyes during Bjerkandera adusta CCBAS 930 cultures.
Biotransformation and toxicity effect of monoanthraquinone dyes during Bjerkandera adusta CCBAS 930 cultures.
Ecotoxicology and environmental safety (2020-01-24)
K Rybczyńska-Tkaczyk, T Korniłłowicz-Kowalska, K A Szychowski, J Gmiński
ABSTRACT
The aim of this study was to evaluate of possibility of biotransformation and toxicity effect of monoanthraquinone dyes in cultures of Bjerkandera adusta CCBAS 930. Phenolic compounds, free radicals, phytotoxicity (Lepidium sativum L.), ecotoxicity (Vibrio fischeri) and cytotoxicity effect were evaluated to determine the toxicity of anthraquinone dyes before and after the treatment with B. adusta CCBAS 930. More than 80% of ABBB and AB129 was removed by biodegradation (decolorization) and biosorption, but biodegradation using oxidoreductases was the main dye removing mechanism. Secondary products toxic to plants and bacteria were formed in B. adusta strain CCBAS 930 cultures, despite efficient decolorization. ABBB and AB129 metabolites increased reactive oxygen species (ROS) production in human fibroblasts, but did not increase LDH release, did not affect the resazurine reduction assay and did not change caspase-9 or caspase-3 activity.
Sigma-Aldrich
3,4-Dimethoxybenzyl alcohol, 96%