Merck
  • Smad4 promotes diabetic nephropathy by modulating glycolysis and OXPHOS.
Smad4 promotes diabetic nephropathy by modulating glycolysis and OXPHOS.
EMBO reports (2020-01-10)
Jinhua Li, Yu Bo Yang Sun, Weiyi Chen, Jinjin Fan, Songhui Li, Xinli Qu, Qikang Chen, Riling Chen, Dajian Zhu, Jinfeng Zhang, Zhuguo Wu, Honggang Chi, Simon Crawford, Viola Oorschot, Victor G Puelles, Peter G Kerr, Yi Ren, Susan K Nilsson, Mark Christian, Huanwen Tang, Wei Chen, John F Bertram, David J Nikolic-Paterson, Xueqing Yu
ABSTRACT
Diabetic nephropathy (DN) is the leading cause of end-stage kidney disease. TGF-β1/Smad3 signalling plays a major pathological role in DN; however, the contribution of Smad4 has not been examined. Smad4 depletion in the kidney using anti-Smad4 locked nucleic acid halted progressive podocyte damage and glomerulosclerosis in mouse type 2 DN, suggesting a pathogenic role of Smad4 in podocytes. Smad4 is upregulated in human and mouse podocytes during DN. Conditional Smad4 deletion in podocytes protects mice from type 2 DN, independent of obesity. Mechanistically, hyperglycaemia induces Smad4 localization to mitochondria in podocytes, resulting in reduced glycolysis and oxidative phosphorylation and increased production of reactive oxygen species. This operates, in part, via direct binding of Smad4 to the glycolytic enzyme PKM2 and reducing the active tetrameric form of PKM2. In addition, Smad4 interacts with ATPIF1, causing a reduction in ATPIF1 degradation. In conclusion, we have discovered a mitochondrial mechanism by which Smad4 causes diabetic podocyte injury.
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, HRP conjugate, Upstate®, from goat
Sigma-Aldrich
Suberic acid bis(N-hydroxysuccinimide ester), ≥95%, powder
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, HRP-conjugate, 1 mg/mL, Upstate®