Merck
  • Morphological and molecular aspects of immobilization-induced muscle atrophy in rats at different stages of postnatal development: the role of autophagy.
Morphological and molecular aspects of immobilization-induced muscle atrophy in rats at different stages of postnatal development: the role of autophagy.
Journal of applied physiology (Bethesda, Md. : 1985) (2016-07-23)
Camila Silva Foresto, Sílvia Paula-Gomes, Wilian Assis Silveira, Flávia Aparecida Graça, Isis do Carmo Kettelhut, Dawit Albieiro Pinheiro Gonçalves, Ana Claudia Mattiello-Sverzut
ABSTRACT
Muscle loss occurs following injury and immobilization in adulthood and childhood, which impairs the rehabilitation process; however, far fewer studies have been conducted analyzing atrophic response in infants. This work investigated first the morphological and molecular mechanisms involved in immobilization-induced atrophy in soleus muscles from rats at different stages of postnatal development [i.e., weanling (WR) and adult (AR) rats] and, second, the role of autophagy in regulating muscle plasticity during immobilization. Hindlimb immobilization for 10 days reduced muscle mass and fiber cross-sectional area, with more pronounced atrophy in WR, and induced slow-to-fast fiber switching. These effects were accompanied by a decrease in markers of protein synthesis and an increase in autophagy. The ubiquitin (Ub)-ligase MuRF1 and the ubiquitinated proteins were upregulated by immobilization in AR while the autolyzed form of μ-calpain was increased in WR. To further explore the role of autophagy in muscle abnormalities, AR were concomitantly immobilized and treated with colchicine, which blocks autophagosome-lysosome fusion. Colchicine-treated immobilized muscles had exacerbated atrophy and presented degenerative features. Despite Igf1/Akt signaling was downregulated in immobilized muscles from both age groups, Foxo1 and 4 phosphorylation was increased in WR. In the same group of animals, Foxo1 acetylation and Foxo1 and 4 content was increased and decreased, respectively. Our data show that muscle disorders induced by 10-day-immobilization occur in both age-dependent and -independent manners, an understanding that may optimize treatment outcomes in infants. We also provide further evidence that the strong inhibition of autophagy may be ineffective for treating muscle atrophy.
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Sigma-Aldrich
Anti-Myosin Antibody, slow muscle, clone NOQ7.5.4D, clone NOQ7.5.4D, Chemicon®, from mouse