Merck
  • ELF3 is an antagonist of oncogenic-signalling-induced expression of EMT-TF ZEB1.
ELF3 is an antagonist of oncogenic-signalling-induced expression of EMT-TF ZEB1.
Cancer biology & therapy (2018-08-28)
D Liu, Y Skomorovska, J Song, E Bowler, R Harris, M Ravasz, S Bai, M Ayati, K Tamai, M Koyuturk, X Yuan, Z Wang, Y Wang, R M Ewing
ABSTRACT
Background: Epithelial-to-mesenchymal transition (EMT) is a key step in the transformation of epithelial cells into migratory and invasive tumour cells. Intricate positive and negative regulatory processes regulate EMT. Many oncogenic signalling pathways can induce EMT, but the specific mechanisms of how this occurs, and how this process is controlled are not fully understood. Methods: RNA-Seq analysis, computational analysis of protein networks and large-scale cancer genomics datasets were used to identify ELF3 as a negative regulator of the expression of EMT markers. Western blotting coupled to siRNA as well as analysis of tumour/normal colorectal cancer panels was used to investigate the expression and function of ELF3. Results: RNA-Seq analysis of colorectal cancer cells expressing mutant and wild-type β-catenin and analysis of colorectal cancer cells expressing inducible mutant RAS showed that ELF3 expression is reduced in response to oncogenic signalling and antagonizes Wnt and RAS oncogenic signalling pathways. Analysis of gene-expression patterns across The Cancer Genome Atlas (TCGA) and protein localization in colorectal cancer tumour panels showed that ELF3 expression is anti-correlated with β-catenin and markers of EMT and correlates with better clinical prognosis. Conclusions: ELF3 is a negative regulator of the EMT transcription factor (EMT-TF) ZEB1 through its function as an antagonist of oncogenic signalling.
Sigma-Aldrich
Anti-ELF3 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution