Merck
  • Dissociating beta-amyloid from alpha 7 nicotinic acetylcholine receptor by a novel therapeutic agent, S 24795, normalizes alpha 7 nicotinic acetylcholine and NMDA receptor function in Alzheimer's disease brain.
Dissociating beta-amyloid from alpha 7 nicotinic acetylcholine receptor by a novel therapeutic agent, S 24795, normalizes alpha 7 nicotinic acetylcholine and NMDA receptor function in Alzheimer's disease brain.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2009-09-04)
Hoau-Yan Wang, Andres Stucky, JingJing Liu, Changpeng Shen, Caryn Trocme-Thibierge, Philippe Morain
ABSTRACT
Alzheimer's disease (AD) is characterized by synaptic dysfunction and cardinal neuropathological features including amyloid plaques and neurofibrillary tangles. Soluble amyloid-beta (Abeta) can suppress synaptic activities by interacting with alpha7 nicotinic acetylcholine receptors (alpha7nAChRs). Here, we show that alpha7nAChR and NMDA glutamatergic receptor (NMDAR) activities are severely compromised in synaptosomes prepared from AD and Abeta(1-42) (Abeta42)-exposed control frontal cortex slices from postmortem tissue. Whereas Abeta(12-28) prevents Abeta42 from binding to alpha7nAChRs, 2-[2-(4-bromophenyl)-2-oxoethyl]-1-methyl pyridinium (S 24795), a novel alpha7nAChR partial agonist, facilitates release of Abeta42 from Abeta42-alpha7nAChR and -Abeta42 complexes. S 24795 interacts with alpha7nAChR and Abeta(15-20) region of the Abeta42 to enable partial recovery of the alpha7nAChR and NMDAR channel function. These findings suggest that the Abeta-alpha7nAChR interaction may be an upstream pathogenic event in AD and demonstrate that some recovery of neuronal channel activities may be achieved in AD brains by removing Abeta from alpha7nAChRs.
Sigma-Aldrich
Anti-NR3A Antibody, Upstate®, from rabbit