- Analysis of the role of ubiquitin-interacting motifs in ubiquitin binding and ubiquitylation.
Analysis of the role of ubiquitin-interacting motifs in ubiquitin binding and ubiquitylation.
The ubiquitin-interacting motif (UIM) is a short peptide motif with the dual function of binding ubiquitin and promoting ubiquitylation. This motif is conserved throughout eukaryotes and is present in numerous proteins involved in a wide variety of cellular processes including endocytosis, protein trafficking, and signal transduction. We previously reported that the UIMs of epsin were both necessary and sufficient for its ubiquitylation. In this study, we found that many, but not all, UIM-containing proteins were ubiquitylated. When expressed as chimeric fusion proteins, most UIMs promoted ubiquitylation of the chimera. In contrast to previous studies, we found that UIMs do not exclusively promote monoubiquitylation but rather a mixture of mono-, multi-, and polyubiquitylation. However, UIM-dependent polyubiquitylation does not lead to degradation of the modified protein. UIMs also bind polyubiquitin chains of varying lengths and to different degrees, and this activity is required for UIM-dependent ubiquitylation. Mutational analysis of the UIM revealed specific amino acids that are important for both polyubiquitin binding and ubiquitin conjugation. Finally we provide evidence that UIM-dependent ubiquitylation inhibits the interaction of UIM-containing proteins with other ubiquitylated cellular proteins. Our results suggest a new model for the ubiquitylation of UIM-containing proteins.