Merck
  • Prolonged residence time of a noncovalent molecular adapter, beta-cyclodextrin, within the lumen of mutant alpha-hemolysin pores.
Prolonged residence time of a noncovalent molecular adapter, beta-cyclodextrin, within the lumen of mutant alpha-hemolysin pores.
The Journal of general physiology (2001-11-07)
L Q Gu, S Cheley, H Bayley
ABSTRACT
Noncovalent molecular adapters, such as cyclodextrins, act as binding sites for channel blockers when lodged in the lumen of the alpha-hemolysin (alphaHL) pore, thereby offering a basis for the detection of a variety of organic molecules with alphaHL as a sensor element. beta-Cyclodextrin (betaCD) resides in the wild-type alphaHL pore for several hundred microseconds. The residence time can be extended to several milliseconds by the manipulation of pH and transmembrane potential. Here, we describe mutant homoheptameric alphaHL pores that are capable of accommodating betaCD for tens of seconds. The mutants were obtained by site-directed mutagenesis at position 113, which is a residue that lies near a constriction in the lumen of the transmembrane beta barrel, and fall into two classes. Members of the tight-binding class, M113D, M113N, M113V, M113H, M113F and M113Y, bind betaCD approximately 10(4)-fold more avidly than the remaining alphaHL pores, including WT-alphaHL. The lower K(d) values of these mutants are dominated by reduced values of k(off). The major effect of the mutations is most likely a remodeling of the binding site for betaCD in the vicinity of position 113. In addition, there is a smaller voltage-sensitive component of the binding, which is also affected by the residue at 113 and may result from transport of the neutral betaCD molecule by electroosmotic flow. The mutant pores for which the dwell time of betaCD is prolonged can serve as improved components for stochastic sensors.