Merck
  • Epigenetic modulation of Wnt signaling contributes to neuropathic pain in rats.
Epigenetic modulation of Wnt signaling contributes to neuropathic pain in rats.
Molecular medicine reports (2015-06-23)
Wei Feng, Ruynag Teng, Yang Zhao, Jie Gao, Haichen Chu
ABSTRACT
Previous studies have demonstrated that the Wnt/β‑catenin signaling pathway is critical to the induction and maintenance of chronic neuropathic pain caused by peripheral inflammation and nerve damage. Emerging evidence from recent studies suggests that epigenetic mechanisms may also be critical to the pathogenesis of chronic pain. The present study aimed to elucidate the epigenetic mechanisms underlying altered Wnt signaling and their involvement in CCI‑induced neuropathic pain in rat sciatic nerves. The results of the present study demonstrated a significant increase in the expression levels of Wnt3a in the dorsal horn of the rats with CCI. In addition, a significant increase in histone H3 acetylation, and a significant decrease in cytosine methylation in the promoter region of Wnt3a was observed in the dorsal horn of the rats with CCI. Intrathecal application of XAV939, which acts as an inhibitor of Wnt signaling, significantly decreased the expression levels of active β‑catenin, and attenuated the rat behavioral responses to thermal and mechanical pain stimuli. These results suggest that the epigenetic upregulation of Wnt3a in the dorsal horn contributes to the maintenance of pain‑induced behavior in rats with CCI.
Sigma-Aldrich
Anti-Beta Catenin Antibody, clone 7F7.2, clone 7F7.2, Upstate®, from mouse