Merck
FTO affects hippocampal function by regulation of BDNF processing.
PloS one (2019-02-08)
André Spychala, Ulrich Rüther
ABSTRACT
Initially, the function of the fat mass and obesity associated (Fto) gene seemed to be primarily the regulation of the body weight. Here we show that loss of Fto results in a hyperactivation of the hypothalamic-pituitary-adrenal (HPA) axis. In consequence, Fto-/- mice display an anxiety-like behavior and impairments in working memory. Furthermore, differentiation of neurons is affected in the hippocampus. As a cause of these impairments we identified a processing defect of the neurotrophin BDNF which is most likely the result of a reduced expression of MMP-9. Therefore, we propose FTO as a possible new target to develop novel approaches for the treatment of diseases associated with hippocampal disorders. In parallel, we also would like to make the point that any anti-obesity therapy via blocking FTO function can have negative effects on the proper function of the hippocampus.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution