Merck
  • An induced pluripotent stem cell line (TRNDi009-C) from a Niemann-Pick disease type A patient carrying a heterozygous p.L302P (c.905 T>C) mutation in the SMPD1 gene.
An induced pluripotent stem cell line (TRNDi009-C) from a Niemann-Pick disease type A patient carrying a heterozygous p.L302P (c.905 T>C) mutation in the SMPD1 gene.
Stem cell research (2019-05-28)
Amanda Baskfield, Rong Li, Jeanette Beers, Jizhong Zou, Chengyu Liu, Wei Zheng
ABSTRACT
Niemann-Pick disease type A (NPA) is a rare autosomal recessive lysosomal storage disease caused by mutations in the SMPD1 gene, which encodes for the protein acid sphingomyelinase. A human induced pluripotent stem cell (iPSC) line was generated from dermal fibroblasts of a 21-fetal-week-old female patient with NPA that has a heterozygous mutation of a p.L302P variant (c.905 T > C) using non-integrating Sendai virus technique. This iPSC line offers a useful resource to study the disease pathophysiology and as a cell-based model for drug development to treat NPA.
Sigma-Aldrich
Milli-Mark® Anti-Nanog-Alexa Fluor 488 Antibody, NT, Milli-Mark®, from rabbit