Merck
  • Nuclear receptors regulate alternative lengthening of telomeres through a novel noncanonical FANCD2 pathway.
Nuclear receptors regulate alternative lengthening of telomeres through a novel noncanonical FANCD2 pathway.
Science advances (2019-10-22)
Mafei Xu, Jun Qin, Leiming Wang, Hui-Ju Lee, Chung-Yang Kao, Dan Liu, Zhou Songyang, Junjie Chen, Ming-Jer Tsai, Sophia Y Tsai
ABSTRACT
Alternative lengthening of telomeres (ALT) is known to use homologous recombination (HR) to replicate telomeric DNA in a telomerase-independent manner. However, the detailed process remains largely undefined. It was reported that nuclear receptors COUP-TFII and TR4 are recruited to the enriched GGGTCA variant repeats embedded within ALT telomeres, implicating nuclear receptors in regulating ALT activity. Here, we identified a function of nuclear receptors in ALT telomere maintenance that involves a direct interaction between COUP-TFII/TR4 and FANCD2, the key protein in the Fanconi anemia (FA) DNA repair pathway. The COUP-TFII/TR4-FANCD2 complex actively induces the DNA damage response by recruiting endonuclease MUS81 and promoting the loading of the PCNA-POLD3 replication complex in ALT telomeres. Furthermore, the COUP-TFII/TR4-mediated ALT telomere pathway does not require the FA core complex or the monoubiquitylation of FANCD2, key steps in the canonical FA pathway. Thus, our findings reveal that COUP-TFII/TR4 regulates ALT telomere maintenance through a novel noncanonical FANCD2 pathway.
Sigma-Aldrich
Anti-phospho-ATM (Ser1981) Antibody, clone 10H11.E12, clone 10H11.E12, Upstate®, from mouse
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Anti-FANCA Antibody, clone 5G9, clone 5G9, from mouse