Merck
  • Foliar persistence and residual activity of four insecticides of different mode of action on the predator Engytatus varians (Hemiptera: Miridae).
Foliar persistence and residual activity of four insecticides of different mode of action on the predator Engytatus varians (Hemiptera: Miridae).
Chemosphere (2019-07-01)
Sinue I Morales, Ana M Martínez, José I Figueroa, Jesús Campos-García, Alberto Gómez-Tagle, Philippe Lobit, Guy Smagghe, Samuel Pineda
ABSTRACT
A greenhouse study was conducted to investigate the degradation kinetics of spinosad, flufenoxuron, dimethoate and imidacloprid in tomato (Solanum lycopersicum L.) foliage and their residual toxicity on Engytatus varians (Distant) (Hemiptera: Miridae), a predator of the tomato psyllid Bactericera cockerelli (Sulcer) (Hemiptera: Triozidae). Insecticides were sprayed at 100% and 50% of their maximum field-registered concentrations (MFRC). Starting 6 h after spraying, leaf samples were taken every 10 d for 40 d and analyzed while E. varians adults were exposed to treated leaves to evaluate residual toxicity. Immediately after application at 100% MFRC, the residue concentrations were 73.34 μg g-1 spinosyn A and 59.2 μg g-1 spinosyn D, 9.21 μg g-1 flufenoxuron, 71.49 μg g-1 dimethoate and 31.74 μg g-1 imidacloprid. At 50% MFRC, initial residue concentrations were between 75% and 90% those at 100% MFRC. The estimated half-life (DT50) of spinosyns A and D, flufenoxuron, and dimethoate was between 34 and 40 d, while that of imidacloprid was 112 d. Flufenoxuron caused no mortality, while mortality due to spinosad was less than 10%, and only during the first 10 d. Mortality caused by either imidacloprid or dimethoate was around 100% up to 10 d after application, then decreased to around 30% after 40 d. Dimethoate toxicity was approximately proportional to residue concentration, while for imidacloprid there was an apparent threshold around 15 μg g-1. These results can be used to establish periods harmless for release of E. varians in the control of B. cockerelli on tomato crops under greenhouse conditions.
Product Description
Supelco
Flufenoxuron, PESTANAL®, analytical standard