Merck
  • Site-Selective Antibody Functionalization via Orthogonally Reactive Arginine and Lysine Residues.
Site-Selective Antibody Functionalization via Orthogonally Reactive Arginine and Lysine Residues.
Cell chemical biology (2019-06-25)
Dobeen Hwang, Napon Nilchan, Alex R Nanna, Xiaohai Li, Michael D Cameron, William R Roush, HaJeung Park, Christoph Rader
ABSTRACT
Homogeneous antibody-drug conjugates (ADCs) that use a highly reactive buried lysine (Lys) residue embedded in a dual variable domain (DVD)-IgG1 format can be assembled with high precision and efficiency under mild conditions. Here we show that replacing the Lys with an arginine (Arg) residue affords an orthogonal ADC assembly that is site-selective and stable. X-ray crystallography confirmed the location of the reactive Arg residue at the bottom of a deep pocket. As the Lys-to-Arg mutation is confined to a single residue in the heavy chain of the DVD-IgG1, heterodimeric assemblies that combine a buried Lys in one arm, a buried Arg in the other arm, and identical light chains, are readily assembled. Furthermore, the orthogonal conjugation chemistry enables the loading of heterodimeric DVD-IgG1s with two different cargos in a one-pot reaction and thus affords a convenient platform for dual-warhead ADCs and other multifaceted antibody conjugates.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture
Supelco
2-[4-(1-Hydroxy-2-methylpropyl)phenyl]propanoic acid, pharmaceutical impurity standard