- PLD1-dependent PKCgamma activation downstream to Src is essential for the development of pathologic retinal neovascularization.
PLD1-dependent PKCgamma activation downstream to Src is essential for the development of pathologic retinal neovascularization.
Vascular endothelial growth factor (VEGF) appears to be an important mediator of pathologic retinal angiogenesis. In understanding the mechanisms of pathologic retinal neovascularization, we found that VEGF activates PLD1 in human retinal microvascular endothelial cells, and this event is dependent on Src. In addition, VEGF activates protein kinase C-gamma (PKCgamma) via Src-dependent PLD1 stimulation. Inhibition of Src, PLD1, or PKCgamma via pharmacologic, dominant negative mutant, or siRNA approaches significantly attenuated VEGF-induced human retinal microvascular endothelial cell migration, proliferation, and tube formation. Hypoxia also induced Src-PLD1-PKCgamma signaling in retina, leading to retinal neovascularization. Furthermore, siRNA-mediated down-regulation of VEGF inhibited hypoxia-induced Src-PLD1-PKCgamma activation and neovascularization. Blockade of Src-PLD1-PKCgamma signaling via the siRNA approach also suppressed hypoxia-induced retinal neovascularization. Thus, these observations demonstrate, for the first time, that Src-dependent PLD1-PKCgamma activation plays an important role in pathologic retinal angiogenesis.