- Pharmacological dissection of K(v)7.1 channels in systemic and pulmonary arteries.
Pharmacological dissection of K(v)7.1 channels in systemic and pulmonary arteries.
The aim of this study was to characterize the functional impact of KCNQ1-encoded voltage-dependent potassium channels (K(v)7.1) in the vasculature. Mesenteric arteries, intrapulmonary arteries and thoracic aortae were isolated from adult rats. K(v)7.1 channel expression was established by fluorescence immunocytochemistry. Wire myography determined functionality of these channels in response to selective blockers and activators. Xenopus oocytes expressing K(v)7.1 channels were used to assess the effectiveness of selective K(v)7.1 channel blockers. K(v)7.1 channels were identified in arterial myocytes by immunocytochemistry. K(v)7.1 blockers HMR1556, L-768,673 (10 ĀµM) and JNJ39490282 (JNJ282; 1 ĀµM) had no contractile effects in arteries, whereas the pan-K(v)7 channel blocker linopirdine (10 ĀµM) evoked robust contractions. Application of two compounds purported to activate K(v)7.1 channels, L-364 373 (R-L3) and mefenamic acid, relaxed mesenteric arteries preconstricted by methoxamine. These responses were reversed by HMR1556 or L-768,673 but not JNJ282. Similar effects were observed in the thoracic aorta and intrapulmonary arteries. In contrast to previous assumptions, K(v)7.1 channels expressed in arterial myocytes are functional ion channels. Although these channels do not appear to contribute to resting vascular tone, K(v)7.1 activators were effective vasorelaxants.