- Gemcitabine Induces Microvesicle Particle Release in a Platelet-Activating Factor-Receptor-Dependent Manner via Modulation of the MAPK Pathway in Pancreatic Cancer Cells.
Gemcitabine Induces Microvesicle Particle Release in a Platelet-Activating Factor-Receptor-Dependent Manner via Modulation of the MAPK Pathway in Pancreatic Cancer Cells.
Studies, including ours, have shown that pro-oxidative stressors, such as chemotherapeutic agents, generate oxidized lipids with agonistic platelet-activating factor (PAF) activity. Importantly, recent reports have implicated that these PAF-agonists are transported extracellularly via microvesicle particles (MVPs). While the role of PAF-receptor (PAF-R) has been implicated in mediating chemotherapy effects, its significance in chemotherapy-mediated MVP release in pancreatic cancer has not been studied. The current studies determined the functional significance of PAF-R in gemcitabine chemotherapy-mediated MVP release in human pancreatic cancer cells. Using PAF-R-expressing (PANC-1) and PAF-R-deficient (Hs766T) cells, we demonstrate that gemcitabine induces MVP release in a PAF-R-dependent manner. Blocking of PAF-R via PAF-R antagonist or inhibition of MVP generation via inhibitor of acid sphingomyelinase (aSMase) enzyme, significantly attenuated gemcitabine-mediated MVP release from PANC-1 cells, however, exerted no effects in Hs766T cells. Notably, MVPs from gemcitabine-treated PANC-1 cells, contained a measurable amount of PAF-agonists. Mechanistically, pretreatment with ERK1/2 or p38 inhibitors significantly abrogated gemcitabine-mediated MVP release, indicating the involvement of mitogen-activated protein kinase (MAPK) pathway in PAF-R-dependent gemcitabine-mediated MVP release. These findings demonstrate the significance of PAF-R in gemcitabine-mediated MVP release, as well as the rationale of evaluating PAF-R targeting agents with gemcitabine against pancreatic cancer.