Merck
  • Prognostic significance of GAD1 overexpression in patients with resected lung adenocarcinoma.
Prognostic significance of GAD1 overexpression in patients with resected lung adenocarcinoma.
Cancer medicine (2019-06-18)
Mitsuhiro Tsuboi, Kazuya Kondo, Kiyoshi Masuda, Shoichiro Tange, Koichiro Kajiura, Tomohiro Kohmoto, Hiromitsu Takizawa, Issei Imoto, Akira Tangoku
ABSTRACT
In a previous genome-wide screening, we identified hypermethylated CpG islands around glutamate decarboxylase 1 (GAD1) in lung adenocarcinoma (LADC). In this study, we aimed to investigate the methylation and expression status of GAD1 and its prognostic value in patients with LADC. GAD1 methylation and mRNA expression status were analyzed using 33 tumorous and paired non-tumorous LADC samples and publicly available datasets. The prognostic value of GAD1 overexpression was investigated using publicly available datasets of mRNA levels and 162 cases of LADC by immunohistochemistry. The methylation and mRNA expression levels of GAD1, each having a positive correlation, were significantly higher in LADC tumors than in paired non-tumorous tissues. LADC patients with higher GAD1 mRNA expression showed significantly poorer prognosis for overall survival in publicly available datasets. Higher immunoreactivity of GAD1 was significantly associated with the pathological stage, pleural invasion, lymph vessel invasion, and poorer prognosis for cancer-specific and disease-free survival. Multivariate analysis revealed that GAD1 protein overexpression is an independent prognosticator for disease-free survival. GAD1 mRNA and protein expression levels were significant prognostic factors in LADC, suggesting that they might be useful biomarkers to stratify patients with worse clinical outcomes after resection.