Merck
  • Changes in lipids composition and metabolism in colorectal cancer: a review.
Changes in lipids composition and metabolism in colorectal cancer: a review.
Lipids in health and disease (2019-01-28)
Alicja Pakiet, Jarosław Kobiela, Piotr Stepnowski, Tomasz Sledzinski, Adriana Mika
ABSTRACT
Altered metabolism of lipids is currently considered a hallmark characteristic of many malignancies, including colorectal cancer (CRC). Lipids are a large group of metabolites that differ in terms of their fatty acid composition. This review summarizes recent evidence, documenting many alterations in the content and composition of fatty acids, polar lipids, oxylipins and triacylglycerols in CRC patients' sera, tumor tissues and adipose tissue. Some of altered lipid molecules may be potential biomarkers of CRC risk, development and progression. Owing to a significant role of many lipids in cancer cell metabolism, some of lipid metabolism pathways may also constitute specific targets for anti-CRC therapy.