- Identification and functional characterization of IRAK-4 in grass carp (Ctenopharyngodon idellus).
Identification and functional characterization of IRAK-4 in grass carp (Ctenopharyngodon idellus).
IL-1R-associated kinase 4 (IRAK4), a central TIR signaling mediator in innate immunity, can initiate a cascade of signaling events and lead to induction of inflammatory target gene expression eventually. In the present study, we cloned and characterized an IRAK4 orthologue from grass carp (Ctenopharyngodon idella). The full length cDNA of CiIRAK4 was 2057 bp with an ORF of 1422 bp encoding a polypeptide of 472 amino acids. Multiple alignments showed that IRAK4s were highly conserved among different species. Phylogenetic tree analysis revealed that CiIRAK4 shared high homologous with zebra fish IRAK4. Expression analysis indicated that CiIRAK4 was widely expressed in all tested tissues. It was significantly up-regulated after treatment with poly I:C, especially obvious in liver and spleen. Also, CiIRAK4 could be induced by poly I:C and LPS in CIK cells. Fluorescence microscopy assays showed that CiIRAK4 localized in the cytoplasm. RNAi-mediated knockdown and overexpression assays indicated that CiIRAK4 might have little effect on NF-kappa B p65 translocation from cytoplasm to nucleus, indicating that CiIRAK4 was dispensable for activation of NF-kappa B p65. In addition, IRAK4 promoted IRF5 nuclear translocation, which has nothing to do with the interaction between IRAK4 and IRF5. It suggested that fish IRAK4 kinase regulated IRF5 activity through indirect ways.