Merck
  • Water compatible imprinted polymer prepared in water for selective solid phase extraction and determination of ciprofloxacin in real samples.
Water compatible imprinted polymer prepared in water for selective solid phase extraction and determination of ciprofloxacin in real samples.
Talanta (2019-05-01)
Guifen Zhu, Guohao Cheng, Peiyun Wang, Wanwan Li, Yicong Wang, Jing Fan
ABSTRACT
A novel water compatible ciprofloxacin imprinted polymer is synthesized in water via a green, non-toxic and environmentally friendly polymerization process. Hydrophilic groups, including anionic chlorine, hydroxyl, and carbonyl oxygen provided by a bifunctional monomer comprising 1-allyl-3-vinylimidazole chloride and 2-hydroxyethyl methacrylate, are introduced into the imprinted material, which allows the polymer to interact strongly with imprinting molecule via hydrogen bonds, electrostatic and π-π dipole interactions in aqueous solution. Rebinding experiments show that the obtained molecularly imprinted polymer (MIP) presents special molecular recognition towards quinolone antibiotics (ciprofloxacin, levofloxacin and pefloxacin mesylate) in aqueous matrices. The adsorption process of ciprofloxacin on MIP and non-imprinted polymer (NIP) substrates involves spontaneous exothermic reactions, and the maximum rebinding capacities of ciprofloxacin on MIP and NIP at 25 °C are 19.96 and 8.86 mg g-1, respectively. The excellent selectivity and hydrophilicity of this imprinted polymer makes it suitable for use as an adsorbent in solid phase extraction. Under the optimized conditions, the presented MIP-SPE protocol exhibits a wide linear range between 0.29 and 1.47 × 105 μg L-1 and has been successfully applied for the separation and enrichment of trace ciprofloxacin in real water, soil and pork samples with satisfactory recoveries of 87.33-102.50%. The proposed study implies the promising prospect of this green and water compatible MIP in highly effective recognition and separation of trace quinolones in complex matrics.