Merck
  • The alkyl-connected 2-amino-6-vinylpurine (AVP) crosslinking agent for improved selectivity to the cytosine base in RNA.
The alkyl-connected 2-amino-6-vinylpurine (AVP) crosslinking agent for improved selectivity to the cytosine base in RNA.
Bioorganic & medicinal chemistry (2010-03-30)
Yosuke Taniguchi, Yusuke Kurose, Takamasa Nishioka, Fumi Nagatsugi, Shigeki Sasaki
ABSTRACT
We have previously reported that the 2-amino-6-vinylpurine (AVP) nucleoside exhibits a highly efficient and selective crosslinking reaction toward cytosine and displayed an improved antisense inhibition in cultured cells. In this study, we further investigated the alkyl-connected AVP nucleoside analogs for more efficient crosslinking to the cytosine base (rC) of the target RNA. We synthesized three AVP analogs which connect the 2-amino-6-vinylpurine unit to the 2'-deoxyribose through a methylene, an ethylene, or a butylene linker. The ODN incorporating the AVP analog with the methylene or the butylene linker showed a slightly higher crosslinking to the target rC of RNA than the original AVP with no linker. In contrast, the AVP with the ethylene linker formed a selective and efficient crosslink to the rC of the target RNA.
Sigma-Aldrich
Vinylboronic anhydride pyridine complex, 95%