Merck
  • Sulphur-containing amino acids are agonists for group 1 metabotropic receptors expressed in clonal RGT cell lines.
Sulphur-containing amino acids are agonists for group 1 metabotropic receptors expressed in clonal RGT cell lines.
Neuropharmacology (1998-07-29)
A E Kingston, J Lowndes, N Evans, B Clark, R Tomlinson, J P Burnett, N G Mayne, S L Cockerham, D Lodge
ABSTRACT
Comparison of the pharmacological effects of a range of sulphur-containing amino acids on human mGluR1alpha and mGluR5a has been undertaken. cDNAs of each mGluR were transfected into a Syrian hamster tumour cell line AV12-664 that was previously transfected with the rat glutamate-aspartate transporter protein (GLAST). The L-isomers of cysteine sulphinic acid (CSA), homocysteine sulphinic acid (HCSA), cysteic acid (CA) and serine-O-sulphate (SOS) stimulated PI hydrolysis in human mGluR1alpha and mGluR5a cells with full agonist effects. D-CSA, the only active D-isomer, was a partial agonist for mGluR5a whereas L-sulphocysteine (S-CYS) showed weak agonist-like effects at high concentrations on both mGluR1alpha and mGluR5a. L-Homocysteic acid was inactive on both mGluR1alpha and mGluR5a cells. Treatment of mGluR cultures with glutamate pyruvate transaminase did not alter the potencies of the S-amino acids on PI hydrolysis responses. Inhibitor constants (Ki) obtained for L-HCSA, L-CSA, L-CA and L-SOS in [3H]glutamate receptor binding studies with mGluR1alpha cells indicated that L-HCSA, L-CSA, L-CA and L-SOS can bind specifically to mGluR1 with L-HCSA showing the highest affinity. These results confirm that certain endogenously produced S-amino acids may interact directly with group 1 mGluRs.
Sigma-Aldrich
L-Cysteic acid monohydrate, ≥99.0% (T)