Merck
  • Cancer Immunotherapy with T Cells Carrying Bispecific Receptors That Mimic Antibodies.
Cancer Immunotherapy with T Cells Carrying Bispecific Receptors That Mimic Antibodies.
Cancer immunology research (2019-03-08)
Sarah Ahn, Jingjing Li, Chuang Sun, Keliang Gao, Koichi Hirabayashi, Hongxia Li, Barbara Savoldo, Rihe Liu, Gianpietro Dotti
ABSTRACT
Tumors are inherently heterogeneous in antigen expression, and escape from immune surveillance due to antigen loss remains one of the limitations of targeted immunotherapy. Despite the clinical use of adoptive therapy with chimeric antigen receptor (CAR)-redirected T cells in lymphoblastic leukemia, treatment failure due to epitope loss occurs. Targeting multiple tumor-associated antigens (TAAs) may thus improve the outcome of CAR-T cell therapies. CARs developed to simultaneously target multiple targets are limited by the large size of each single-chain variable fragment and compromised protein folding when several single chains are linearly assembled. Here, we describe single-domain antibody mimics that function within CAR parameters but form a very compact structure. We show that antibody mimics targeting EGFR and HER2 of the ErbB receptor tyrosine kinase family can be assembled into receptor molecules, which we call antibody mimic receptors (amR). These amR can redirect T cells to recognize two different epitopes of the same antigen or two different TAAs in vitro and in vivo.