Merck
  • Ethyl rosmarinate inhibits lipopolysaccharide-induced nitric oxide and prostaglandin E2 production in alveolar macrophages.
Ethyl rosmarinate inhibits lipopolysaccharide-induced nitric oxide and prostaglandin E2 production in alveolar macrophages.
European journal of pharmacology (2018-02-03)
Hathairat Thammason, Pichit Khetkam, Wachirachai Pabuprapap, Apichart Suksamrarn, Duangkamol Kunthalert
ABSTRACT
In this study, a series of rosmarinic acid and analogs were investigated for their anti-inflammatory potential against LPS-induced alveolar macrophages (MH-S). Our results showed that, among the test compounds, ethyl rosmarinate (3) exhibited the most potent inhibitory effect on NO production in LPS-induced MH-S cells, with low cytotoxicity. Compound 3 exhibited remarkable inhibition of the production of PGE2 in LPS-induced MH-S cells. The inhibitory potency of compound 3 against LPS-induced NO and PGE2 release was approximately two-fold higher than that of dexamethasone. Compound 3 significantly decreased the mRNA and protein expression of iNOS and COX-2 and suppressed p65 expression in the nucleus in LPS-induced MH-S cells. These results suggested that compound 3 inhibited NO and PGE2 production, at least in part, through the down-regulation of NF-κB activation. Analysis of structure-activity relationship revealed that the free carboxylic group did not contribute to inhibitory activity and that the alkyl group of the corresponding alkyl ester analogs produced a strong inhibitory effect. We concluded that compound 3, a structurally modified rosmarinic acid, possessed potent inhibitory activity against lung inflammation, which strongly supported the development of this compound as a novel therapeutic agent for the treatment of macrophage-mediated lung inflammatory diseases, such as COPD.
Sigma-Aldrich
Ethyl rosmarinate, ≥98% (HPLC)