Merck
  • Enantioselective Aluminum-Free Alkene Hydroarylations through C-H Activation by a Chiral Nickel/JoSPOphos Manifold.
Enantioselective Aluminum-Free Alkene Hydroarylations through C-H Activation by a Chiral Nickel/JoSPOphos Manifold.
Angewandte Chemie (International ed. in English) (2018-12-06)
Joachim Loup, Valentin Müller, Debasish Ghorai, Lutz Ackermann
ABSTRACT
Highly enantioselective nickel-catalyzed alkene endo-hydroarylations were accomplished with full selectivity by organometallic C-H activation. The asymmetric assembly of chiral six-membered scaffolds proved viable in the absence of pyrophoric organoaluminum reagents within an unprecedented nickel/JoSPOphos manifold.