Merck
  • Characterization of a nonchymotrypsin-like endopeptidase from anterior pituitary that hydrolyzes luteining hormone-releasing hormone at the tyrosyl-glycine and histidyl-tryptophan bonds.
Characterization of a nonchymotrypsin-like endopeptidase from anterior pituitary that hydrolyzes luteining hormone-releasing hormone at the tyrosyl-glycine and histidyl-tryptophan bonds.
Biochemistry (1980-06-24)
B Horsthemke, K Bauer
ABSTRACT
A neutral endopeptidase which degrades luteinizing hormone-releasing hormone (LH-RH, <GLu-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-GLy-NH2) has been purified 900-fold from extracts of bovine anterior pituitary. This Ca2+-independent enzyme of 83 000 molecular weight (as estimated by gel filtration) cleaves LH-RH (KM = 180 microM) at the Tyr5-Gly6-His2-Trp3 bonds. Its activity is inhibited by the SH-reactive agents N-ethylmaleimide and p-(chloromercuri)benzoate but not by the OH-reactive agent diisopropyl fluorophosphate. Hydrolysis of the fluorogenic chymotrypsin substrate glutarkyl-Gly-Gly-Phe-beta-naphthylamide by this endopeptidase could not be detected. These properties differentiate the endopeptidase from chymotrypsin and from a glutaryl-Gly-Gly-Phe-beta-naphthylamide hydrolyzing activity of high molecular weight, which has been isolated from the same tissue and also hydrolyzes internal bonds of LH-RH.
Sigma-Aldrich
N-Succinyl-Phe-Gly-Leu-β-naphthylamide