Merck
  • Analysis of the role of the PAC1 receptor in neutrophil recruitment, acute-phase response, and nitric oxide production in septic shock.
Analysis of the role of the PAC1 receptor in neutrophil recruitment, acute-phase response, and nitric oxide production in septic shock.
Journal of leukocyte biology (2005-01-22)
Carmen Martínez, Yasmina Juarranz, Catalina Abad, Alicia Arranz, Begoña G Miguel, Florencia Rosignoli, Javier Leceta, Rosa P Gomariz
ABSTRACT
Infections caused by Gram-negative bacteria constitute one of the major causes of septic shock, which results from the inability of the immune system to limit bacterial spread during the ongoing infection. In the last decade, it has been demonstrated that vasoactive intestinal peptide (VIP) and pituitary adenylate cyclase-activating polypeptide (PACAP) are two endogenous immunopeptides, which together with three G protein-coupled receptors (VPAC1, VPAC2, and PAC1) exert a significant, therapeutic effect attenuating the deleterious consequences of septic shock by balancing pro- and anti-inflammatory factors. We have recently shown PAC1 receptor involvement in vivo as an anti-inflammatory receptor, at least in part, by attenuating lipopolysaccharide-induced production of proinflammatory interleukin-6. The present study deepens in the protective role of PAC1 receptor in septic shock, elucidating its involvement in the modulation of neutrophil recruitment and in the expression of different molecular sensors such as intercellular adhesion molecule-1, vascular cell adhesion molecule-1, fibrinogen, serum amyloid A, and nitric oxide as important, systemic players of the development of septic shock. Our results, using a mice deficient in PAC1 and a PAC1 antagonist, show that VIP and PACAP as well as the PAC1 receptor are involved in neutrophil recruitment in different target organs, in adhesion molecules expression, and in coagulation-related molecule fibrinogen synthesis. Thus, this study provides some important insights with respect to the involvement of PAC1 into the complexities of sepsis and represents an advantage for the design of more specific drugs complementing standard intensive care therapy in severe sepsis, confirming VIP and PACAP as candidates for multitarget therapy of septic shock.
Sigma-Aldrich
ChemiSCREEN Human PAC1-long Receptor Membrane Preparation, Human PAC1 long isoform / PACAP GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.