Merck
  • Purinergic control of T cell activation by ATP released through pannexin-1 hemichannels.
Purinergic control of T cell activation by ATP released through pannexin-1 hemichannels.
Science signaling (2008-10-02)
Ursula Schenk, Astrid M Westendorf, Enrico Radaelli, Anna Casati, Micol Ferro, Marta Fumagalli, Claudia Verderio, Jan Buer, Eugenio Scanziani, Fabio Grassi
ABSTRACT
T cell receptor (TCR) stimulation results in the influx of Ca(2+), which is buffered by mitochondria and promotes adenosine triphosphate (ATP) synthesis. We found that ATP released from activated T cells through pannexin-1 hemichannels activated purinergic P2X receptors (P2XRs) to sustain mitogen-activated protein kinase (MAPK) signaling. P2XR antagonists, such as oxidized ATP (oATP), blunted MAPK activation in stimulated T cells, but did not affect the nuclear translocation of the transcription factor nuclear factor of activated T cells, thus promoting T cell anergy. In vivo administration of oATP blocked the onset of diabetes mediated by anti-islet TCR transgenic T cells and impaired the development of colitogenic T cells in inflammatory bowel disease. Thus, pharmacological inhibition of ATP release and signaling could be beneficial in treating T cell-mediated inflammatory diseases.
Sigma-Aldrich
Adenosine 5′-triphosphate, periodate oxidized sodium salt, ≥97%