- Structure of a halophilic nucleoside diphosphate kinase from Halobacterium salinarum.
Structure of a halophilic nucleoside diphosphate kinase from Halobacterium salinarum.
FEBS letters (2005-11-19)
Hüseyin Besir, Kornelius Zeth, Andreas Bracher, Ursula Heider, Matsujiro Ishibashi, Masao Tokunaga, Dieter Oesterhelt
PMID16293253
ABSTRACT
Nucleoside diphosphate kinase from the halophilic archaeon Halobacterium salinarum was crystallized in a free state and a substrate-bound form with CDP. The structures were solved to a resolution of 2.35 and 2.2A, respectively. Crystals with the apo-form were obtained with His6-tagged enzyme, whereas the untagged form was used for co-crystallization with the nucleotide. Crosslinking under different salt and pH conditions revealed a stronger oligomerization tendency for the tagged protein at low and high salt concentrations. The influence of the His6-tag on the halophilic nature of the enzyme is discussed on the basis of the observed structural properties.