Merck
  • Methyl de-esterification as a major factor regulating the extent of pectin depolymerization during fruit ripening: a comparison of the action of avocado (Persea americana) and tomato (Lycopersicon esculentum) polygalacturonases.
Methyl de-esterification as a major factor regulating the extent of pectin depolymerization during fruit ripening: a comparison of the action of avocado (Persea americana) and tomato (Lycopersicon esculentum) polygalacturonases.
Journal of plant physiology (2003-07-23)
Kazuyuki Wakabayashi, Takayuki Hoson, Donald J Huber
ABSTRACT
Pectinmethylesterase (PME, EC 3.2.1.11) and polygalacturonase (PG, EC 3.2.1.15) are known to operate in tandem to degrade methylesterified polyuronides. In this study, PGs purified from tomato and avocado fruit were compared in terms of their capacity to hydrolyze water-soluble polyuronides from avocado before and following enzymic or chemical de-esterification. When assayed using polygalacturonic acid or polyuronides from avocado fruit, the activity of PG from tomato fruit was 3-4 times higher than that from avocado fruit. High molecular mass, low methylesterified (33%) water-soluble polyuronides (WSP) from pre-ripe avocado fruit (day 0) were partially depolymerized upon incubation with purified avocado and tomato PGs. In contrast, middle molecular mass, highly methylesterified (74%) WSP from day 2 fruit were largely resistant to the action of both PGs. PME or weak alkali treatment of highly methylesterified WSP decreased the methylesterification values to 11 and 4.5%, respectively. Treatment of de-esterified WSP with either avocado or tomato PGs caused extensive molecular mass downshifts, paralleling those observed during avocado fruit ripening. Although PME and PG are found in many fruits, the pattern of depolymerization of native polyuronides indicates that the degree of cooperativity between these enzymes in vivo differs dramatically among fruits. The contribution of PME to patterns of polyuronide depolymerization observed during ripening compared with physically compromised fruit tissues is discussed.